Know your labware is autoclaved with this autoclave tape. Cole-Parmer™ Autoclave Tape displays a dark colored diagonal after being autoclaved at 121°C for 15 minutes. Adhesive backing sticks to glass, cloth or metal.
Have Questions?
Catalog Number
Dimensions (L x W)
FKS11883282
12 m x 25 mm
Catalog number FKS11883282
Price (MYR)
297.00
EA
Add to cart
Dimensions (L x W):
12 m x 25 mm
Price (MYR)
297.00
EA
Add to cart
Crepe tape peels from surfaces easily and leaves no residue.
Specifications
Dimensions (L x W)12 m x 25 mm
Temperature (Metric)121°C
FeaturesCrepe tape displays a dark colored diagonal after being autoclaved at 250°F (121°C) for 15 min.