Neoprene Tubing
Neoprene Tubing
Actual product may vary
Neoprene Tubing
Used in flow lines for fluids and gases in a variety of applications.
Have Questions?
Catalog NumberDescription
FKS118754928 x 10mm
Catalog number FKS11875492
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
8 x 10mm
  • Resistant to heat, ozone, petroleum oils and hydrocarbon solvents
  • Temperature range: -30°C to 100°C
Specifications
Description8 x 10mm
MaterialNeoprene
Overall Length (Metric)10 m
Unit SizeEA