Check Order Status
Plain Glass Plates
Actual product may vary
Plain Glass Plates
Accessory for Fisherbrand Vertical Electrophoresis Systems.
Have Questions?
Catalog Number
Thickness (Metric)
FKS11847643
2 mm
Catalog number
FKS11847643
Price (MYR)
229.00
PK
Add to cart
Thickness (Metric):
2 mm
Price (MYR)
229.00
PK
Add to cart
Accessory for Fisherbrand Vertical Electrophoresis Systems
Specifications
For Use With (Equipment)
Vertical Gel Electrophoresis System
Type
Plain Glass Plate
Gel Dimensions
10 x 10 cm
Quantity
2/Pk.
Thickness (Metric)
2 mm