Glassine Weighing Papers
Glassine Weighing Papers
Actual product may vary
Fisherbrand
Glassine Weighing Papers
Ensure complete sample transfer with glassine weighing paper. Cole Parmer™ Glassine Weighing Paper are moisture-resistant and nonabsorbent with a smooth glassine surface.
Have Questions?
Catalog NumberDimensions (L x W)
FKS118452127.6 x 7.6 cm
Catalog number FKS11845212
Price (MYR)
409.00
PK
Add to cart
Dimensions (L x W):
7.6 x 7.6 cm
Price (MYR)
409.00
PK
Add to cart
Ensure complete sample transfer with glassine weighing paper. Cole Parmer™ Glassine Weighing Paper are moisture-resistant and nonabsorbent with a smooth glassine surface.
  • Keep balance pans clean
  • Disposable
  • Available in three sizes
Specifications
Dimensions (L x W)7.6 x 7.6 cm
DisposableSingle-use
For Use With (Application)Keep balance pans clean
Length (English)3 in.
Length (Metric)7.6 cm
MaterialPaper
Width (English)3 in.
Width (Metric)7.6 cm
Product TypeWeighing Paper
ShapeSquare
Unit SizePK