Model 80 Cooled Incubator, 76 L, Pre-coated Aluminum
Model 80 Cooled Incubator, 76 L, Pre-coated Aluminum
Actual product may vary
Model 80 Cooled Incubator, 76 L, Pre-coated Aluminum
Temperature Range: -10°C to +50°C
Have Questions?
Catalog NumberCapacity (Metric)
FKS1183341076 L
Catalog number FKS11833410
Price (MYR)
71,294.00
EA
Add to cart
Capacity (Metric):
76 L
Price (MYR)
71,294.00
EA
Add to cart
  • Range of cooled incubators with a white stove enamel steel exterior, foam insulation and a white pre-coated aluminum interior
  • Temperature range: -10°C to 50°C
  • Digital control
  • Fan assisted air circulation
  • Full P. I. D. heating control
  • Self-evaporating condensate drip tray
  • White plastic coated wire shelves
  • RS232 interface
Specifications
Capacity (Metric)76 L
Chamber MaterialPre-coated Aluminum
MaterialStove Enameled Mild Steel
No. of Shelves2
Temperature Range (Metric)-10°C to +50°C
TypeRefrigerated Incubator
Includes2 trays
Model80
Height (Metric) Interior40 cm
Depth (Metric) Interior49 cm
Weight (Metric)40 kg
Unit SizeEA