Scintillation Vial Rack
Scintillation Vial Rack
Actual product may vary
Scintillation Vial Rack
For start to finish sample preparation
Have Questions?
Catalog NumberHolds
FKS11833182Vials
Catalog number FKS11833182
Price (MYR)
795.00
EA
Add to cart
Holds:
Vials
Price (MYR)
795.00
EA
Add to cart
  • Holds 24 scintillation vials of 30 mm diameter for storage, handling and sample preparation
  • Rugged, stable design molded from corrosion-resistant polypropylene
  • Autoclavable
  • Shelf height 4.4 cm (1.75 in.)
Specifications
Product TypeVial Rack
Array8 x 3
AutoclavableAutoclavable
ColorWhite
HoldsVials
MaterialPolypropylene
Positions24
ShapeRectangular
Shelves2 Tiers
StackableNo
SterilityNon-sterile
For Use With (Equipment)30mm dia. Scintillation vials
QuantityPk. of 1
Dimensions (L x W x H)11.75 x 5 x 3.75 in. (29.8 x 12.7 x 9.5cm)
Length (Metric)29.8 cm
Length (English)11.75 in.
Width (Metric)12.7 cm
Width (English)5 in.
Height (Metric)9.5 cm
Height (English)3.75 in.
Diameter (Metric) Inner30 mm
Unit SizeEA