HDPE, Jerry Can (Non-Sterile)
HDPE, Jerry Can (Non-Sterile)
Actual product may vary
Fisherbrand
HDPE, Jerry Can (Non-Sterile)
For Use With: For environmental, industrial uses; Closure Material: LDPE
Have Questions?
Catalog number FKS11826840
Price (MYR)
0.00
PK
Add to cart
Price (MYR)
0.00
PK
Add to cart
  • 2.5L size has molded graduations, a bowed neck and a red leakproof cap with an impervious seal
  • 5L size has a red security cap without a seal
  • 10L size has a leakproof cap with an impervious seal
Specifications
Capacity (English)0.7 gal.
Capacity (Metric)2.5 L
Certifications/ComplianceEC N°1935/2004
Closure ColorRed
Closure MaterialLDPE
Closure TypeScrew Cap with Wad
DescriptionNon-sterile
Diameter (Metric) Inner Neck26 mm
Dimensions (L x W x H)220 x 160 x 105mm
For Use With (Application)For environmental, industrial uses
GraduationsYes
Height (Metric) With Closure105 mm
IncludesLeakproof cap with an impervious seal
Length (Metric)220 mm
MaterialHDPE
Product TypeJerry Can
Width (Metric)160 mm
Unit SizePK