Eutech™ Temperature Electrode PH5TEMB01P
Eutech™ Temperature Electrode PH5TEMB01P
Actual product may vary
Eutech™ Temperature Electrode PH5TEMB01P
Eutech™ offers a selection of temperature meters and electrodes for use in various educational, laboratory and industrial applications.
Have Questions?
Catalog NumberConnector Types
FKS11816332Phono Plug
Catalog number FKS11816332
Price (MYR)
875.00
EA
Add to cart
Connector Types:
Phono Plug
Price (MYR)
875.00
EA
Add to cart
Eutech™ offers a selection of temperature meters and electrodes for use in various educational, laboratory and industrial applications. The Temperature Electrode PH5TEMB01P is a versatile electrode made of stainless steel and has a temeprature range from 0 to 100 ºC.
Features:
  • Parameter: Temperature
  • Temperature Range: 0 to 100 °C
  • Material: Stainless steel 304
  • Dimensions (Shaft): 84 x 3mm
  • Dimensions (Handle): 85 x 12mm
  • Cable Length: 1m
  • Connector: 2.5mm phono plug
  • Used With: pH 110 / pH 11 / Ion 6+ / pH 6+ / pH 5+
Specifications
Temperature (Metric) Operating0°C to 100°C
TypeTemperature Electrode
Connector TypesPhono Plug
Unit SizeEA