Check Order Status
Stainless Steel Spatulas
Actual product may vary
Stainless Steel Spatulas
Manufactured from stainless steel. Bochem™ Stainless Steel Spatulas have a wooden handle.
Have Questions?
Catalog Number
Length (Metric)
Length (Metric) Blade
FKS11805061
250 mm
150 mm
Catalog number
FKS11805061
Price (MYR)
632.00
PK
Add to cart
Length (Metric):
250 mm
Length (Metric) Blade:
150 mm
Price (MYR)
632.00
PK
Add to cart
Wood handle
Manufactured from stainless steel
Specifications
Quantity
1 Ea.
Product Type
Spatula
Length (Metric) Blade
150 mm
Width (Metric)
22 mm
Width (Metric) Blade
14 mm
Blade Material
Stainless Steel
Diameter (Metric) Outer
1mm
Dimensions (L x W)
250 x 22 mm
Handle Material
Wood
Material
Stainless Steel
Length (Metric)
250 mm