Check Order Status
Cardboard Cryobox Inserts, 136mm
Actual product may vary
Cardboard Cryobox Inserts, 136mm
Combine these inserts with Cryoboxes for flexible cell storage and sample handling. Fisherbrand™ Cardboard Cryobox Inserts feature ultra-low temperature resistant cardboard with water repellent finish for sample storage.
Have Questions?
Catalog Number
Height (Metric)
Array
FKS11802533
20mm
8 x 8
Catalog number
FKS11802533
Price (MYR)
224.00
PK
Add to cart
Height (Metric):
20mm
Array:
8 x 8
Price (MYR)
224.00
PK
Add to cart
Ultra low temperature-resistant cardboard
For sample storage
Temperature-resistant down to -80°C
Water-repellent finish
Specifications
Quantity
10 Pack
Material
Cardboard
Product Type
Grid Insert
For Use With (Application)
Sample storage
Holds
Sample storage tubes
Height (Metric)
20mm
Width (Metric)
136mm
Length (Metric)
136mm
Array
8 x 8