Grade 13 Ashless Medium Filtering Quantitative Filter Paper
Grade 13 Ashless Medium Filtering Quantitative Filter Paper
Actual product may vary
Fisherbrand
Grade 13 Ashless Medium Filtering Quantitative Filter Paper
Used for fine precipitates such as magnesium ammonium phosphate and calcium oxalate.
Have Questions?
Catalog NumberFormatDiameter (Metric)
FKS11798953Disc110 mm
Catalog number FKS11798953
Price (MYR)
349.00
PK
Add to cart
Format:
Disc
Diameter (Metric):
110 mm
Price (MYR)
349.00
PK
Add to cart
  • Medium pores filter paper
  • Made with 100% cotton linter fibers
  • Washed with acid and cleaned with demineralized water to obtain high purity
  • Ash content is 0.007%
  • Applications: Chemical, food, beverage industries and atmospheric analysis
Specifications
DescriptionAsh Content: 0.007%
Filter TypeAshless
FormatDisc
MaterialCotton Linters
Particle Retention5 to 8 μm
Pore Size5 to 8 μm
Filtration Speed50 sec.
For Use With (Application)For fine precipitates such as magnesium ammonium phosphate, calcium oxalate, magnesium ammonium arsenate, coarser forms of barium sulphate
Grade13
Quantity100/Pk.
Thickness (Metric)0.16 mm
TypeQuantitative Filter Paper
Diameter (Metric)110 mm
Unit SizePK