HDPE, Jerry Can with Safety Cap (Non-Sterile)
HDPE, Jerry Can with Safety Cap (Non-Sterile)
Actual product may vary
Fisherbrand
HDPE, Jerry Can with Safety Cap (Non-Sterile)
For Use With: For environmental, industrial uses; Closure Material: HDPE
Have Questions?
Catalog number FKS11784624
Price (MYR)
-
  • 2.5L size has molded graduations, a bowed neck and a red leakproof cap with an impervious seal
  • 5L size has a red security cap without a seal
  • 10L size has a leakproof cap with an impervious seal
Specifications
Capacity (English)1.3 gal.
Capacity (Metric)5 L
Certifications/ComplianceEC N°1935/2004
Closure ColorRed
Closure MaterialHDPE
Closure TypeTamper-evident Screw Cap with Wad
DescriptionNon-sterile
Diameter (Metric) Inner Neck32 mm
For Use With (Application)For environmental, industrial uses
IncludesSecurity cap without seal
MaterialHDPE
Product TypeJerry Can
Unit SizePK