Rectangular Retort Stand Bases
Rectangular Retort Stand Bases
Actual product may vary
Fisherbrand
Rectangular Retort Stand Bases
An excellent option for supporting retort stands in the laboratory. R&L Enterprises™ Rectangular Retort Stand Bases are manufactured from white acrylic finished steel.
Have Questions?
Catalog NumberLength (Metric) Base
FKS11779593160 mm
Catalog number FKS11779593
Price (MYR)
235.00
EA
Add to cart
Length (Metric) Base:
160 mm
Price (MYR)
235.00
EA
Add to cart
  • Without rod
  • Fitted with rubber feet
  • Hole threaded M10 at the center of one short side
Specifications
MaterialStainless Steel
Length (Metric) Base160 mm
Width (Metric) Base100 mm
IncludesWith Rubber Feet
Product TypeTripod Plate