Safety Bulb Pipette Filler
Safety Bulb Pipette Filler
Actual product may vary
Fisherbrand
Safety Bulb Pipette Filler
Fill any standard size pipet without the risk of cracking or chipping. Fisherbrand™ Safety Bulb Pipet Filler uses an ABS attachment with a silicone adapter that seals around pipets without twisting. Comes with an elastic band for convenient hanging on bottles or wrist.
Have Questions?
Catalog number FKS11779165
Price (MYR)
225.00
PK
Add to cart
Price (MYR)
225.00
PK
Add to cart
Uses an ABS attachment with a silicone adapter that seals around pipets without twisting. Includes an elastic band for convenient hanging on bottles or wrist. Allows you to fill any standard size pipet without the risk of cracking or chipping.
  • Pipette does not contact hard plastic, which reduces the risk of cracking and chipping
  • 15 mL capacity, 106 mm L x 41 mm OD (4.5 to 6.5 mm hole dia.)
  • Elastic band for hanging on reagent bottle or wrist
  • Filler fits standard size pipettes
Specifications
Compatible Tips4.5 to 6.5 mm
ColorRed
For Use WithPipette
MaterialNatural Rubber, ABS
ModelBulb
Number of Channels1
TypePipette Filler
Volume (Metric)15 mL
For Use With (Equipment)Standard Size Pipettes
IncludesHandy elastic band for hanging on wrist or reagent bottle
Quantity3 Pk.
Length (English)4.21 in.
Length (Metric)107 mm
Diameter (English) Opening0.2 to 0.3 in.
Diameter (English) Outer1.6 in.
Diameter (Metric) Opening4.5 to 6.5 mm
Diameter (Metric) Outer41 mm
Unit SizePK