Externally and Internally Threaded Cryogenic Storage Vials
Externally and Internally Threaded Cryogenic Storage Vials
Actual product may vary
Externally and Internally Threaded Cryogenic Storage Vials
Choice of externally threaded vials with HDPE closures for aseptic technique, or internally threaded vials featuring polypropylene closures with silicone gasket for tight seal
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Thread StyleClosure Material
FKS117779391.2 mLExternalHDPE
FIS10-500-262 mLExternalHDPE
Catalog number FKS11777939
Price (MYR)
296.00
PK
Add to cart
Volume (Metric):
1.2 mL
Thread Style:
External
Closure Material:
HDPE
Price (MYR)
296.00
PK
Add to cart
Choice of externally threaded vials with HDPE closures for aseptic technique, or internally threaded vials featuring polypropylene closures with silicone gasket for tight seal
  • Sterile; not autoclavable
  • Noncytotoxic, nonpyrogenic
  • Large white writing area
  • Graduations in 0.5mL increments
  • Use with Fisherbrand Color Coders for easier sample identification
Externally threaded
  • Conical-style bottom
  • High density polyethylene
  • Self-standing
Internally threaded
  • With silicone gasket
  • Polypropylene
  • Conical- or round-style bottoms with a star-style foot
Vials are for research use only; not for in vitro diagnosis or parenterals.
Suitable for storage of biological materials in mechanical freezers or in the vapor phase of liquid nitrogen
Specifications
Diameter (English) Outer0.53 in.
Diameter (Metric) Outer13.5 mm
MaterialPolypropylene
SterilitySterile
Volume (Metric)1.2 mL
For Use With (Application)For storage of biological materials in mechanical freezers or in the vapor phase of liquid Nitrogen
Height (English)1.49 in.
No. per Case10
No. per Pack100/Pk.
StyleConical
Thread StyleExternal
Closure MaterialHDPE
AutoclavableNot Autoclavable
Closure TypeScrew Cap
Height (Metric)38 mm
Unit SizePK