Interlocking Four-Way Tube Racks
Interlocking Four-Way Tube Racks
Actual product may vary
Fisherbrand
Interlocking Four-Way Tube Racks
Customize your tube rack. Fisherbrand™ Interlocking Four-Way Tube Racks employ an ingenious system of interlocking, 4-way racks. Rotate racks to accommodate 50 mL, 15 mL, 1.5 mL or 0.5 mL tubes, then link them together for whatever size rack you need. Made of sturdy, autoclavable polypropylene.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS11770045Blue
FIS03-448-12Blue
FIS03-448-17Assorted
Catalog number FKS11770045
Price (MYR)
431.00
PK
Add to cart
Color:
Blue
Price (MYR)
431.00
PK
Add to cart
Customize your tube rack. Fisherbrand™ Interlocking Four-Way Tube Racks employ an ingenious system of interlocking, 4-way racks. Rotate racks to accommodate 50mL, 15mL, 1.5mL or 0.5mL tubes, then link them together for whatever size rack you need. Made of sturdy, autoclavable polypropylene.
Create a customized tube rack: rotate and connect.
Rotate each rack for the tube size you need. Each rack can accommodate:
  • 4 x 50 mL conical tubes (Well Dia: 30 mm),
  • 12 x 15 mL conical tubes (Well Dia: 17.5 mm),
  • 32 x 1.5 mL microtubes (Well Dia: 11.6 mm), or
  • 32 x 0.5 mL microtubes (Well Dia: 8.2 mm).
Connect racks to increase the capacity of your tube rack:
  • Unique system of tabs and slots that can link together
  • Create as large a rack as you need for your benchtop applications
  • Available in a range of colors for fun color-coding and easy identification
Specifications
Product Type4-way Microtube Rack
AutoclavableAutoclavable
ColorBlue
HoldsTubes (4 x 50 mL Conical, 12 x 15 mL Conical, 32 x 1.5 mL Microtubes or 32 x 0.5 mL Microtubes)
MaterialPolypropylene
Positions4, 12, 32
ShapeRectangular
SterilityNon-sterile
For Use With (Equipment)Tubes
Dimensions (L x W x H)17.4 x 9.5 x 5.2 cm
Length (Metric)17.4 cm
Length (English)6.9 in.
Width (Metric)9.5 cm
Width (English)3.7 in.
Height (Metric)5.2 cm
Height (English)2 in.
Unit SizePK