Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Actual product may vary
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Fisherbrand
Easy-Squeeze Wash Bottles, 6/Cs.
Easy to fill and ready to use
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorChemical Label
FKS11755243AssortedAssorted Solvents
FIS02-897-6YellowIsopropanol
Catalog number FKS11755243
Price (MYR)
649.00
PK
Add to cart
Color:
Assorted
Chemical Label:
Assorted Solvents
Price (MYR)
649.00
PK
Add to cart
Easy to fill and ready to use
  • Wide mouth
  • Translucent and sputter-free
  • With easy-to-read color-coded silk-screened labels
  • Tips can be cut back to increase flow
  • When not in use, bottles relieve internal pressure without dripping
Label information includes:
  • Chemical name
  • National Fire Protection Association (NFPA) codes and symbols
  • Chemical Abstracts Service (CAS) number
  • Hazard codes
  • Laboratory protective equipment information
  • Target organ information
  • Recommended fill level
  • LDPE recycling code
  • Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement
Not recommended for use above 60°C (140°F).
Specifications
Capacity17 oz., 500mL
Capacity (Metric)500 mL
Capacity (English)17 oz.
ColorAssorted
Diameter (English)2.99 in.
MaterialLDPE
Certifications/ComplianceISO 13485:2003
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthWide
ShapeCylindrical
Height (Metric)173 mm
Product TypeWash Bottle
Height (English)6.81 in.
Chemical LabelAssorted Solvents
Closure MaterialPolypropylene
Closure Size38-415 mm
Diameter (Metric)76 mm
Closure TypePolypropylene Closure
Through the Closure DispensingNo
Unit SizePK