96-Well Reversible Microtube Racks
96-Well Reversible Microtube Racks
Actual product may vary
96-Well Reversible Microtube Racks
96-Well Reversible Microtube Racks
Fisherbrand
96-Well Reversible Microtube Racks
Flip the rack to store a different microtube size. 96-Well Reversible Microtube Racks include easy-to-grasp tabs for secure handling and storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS11738174Natural
FKS11728174Blue, Green, Purple, Yellow, Orange
Catalog number FKS11738174
Price (MYR)
722.00
PK
Add to cart
Color:
Natural
Price (MYR)
722.00
PK
Add to cart
  • Made of polypropylene
  • Standard 8 x 12 array
  • Tabs on each end of rack snap lid into place
  • 0.5 mL wells separated by molded-in grid lines, making them easier to see
  • Raised rings around the 1.5 to 2.0 mL wells make tubes easier to remove
  • Autoclavable
Specifications
IncludesRaised Rings, Lids
Product Type2-in-1 Microcentrifuge Tube Rack
Closure IncludedYes
Array8 x 12
AutoclavableAutoclavable
ColorNatural
HoldsMicrotubes (0.5 mL), Microcentrifuge Tubes (1.5 to 2 mL)
MaterialPolypropylene
Positions96/Side
ShapeRectangular, Rounded Corners
ShelvesNone
StackableYes
SterilityNon-sterile
For Use With (Equipment)1.5 to 2 mL and 0.5 mL Centrifuge Tubes
Quantity5 Pk.
Dimensions (L x W x H)246 x 121 x 50 mm
Length (Metric)246 mm
Length (English)9.7 in.
Width (Metric)121 mm
Width (English)4.8 in.
Height (Metric)50 mm
Height (English)2 in.
Diameter (Metric) Well11 mm, 7.6 mm
Unit SizePK