Class A Volumetric Flasks, Amber Glass
Class A Volumetric Flasks, Amber Glass
Actual product may vary
Class A Volumetric Flasks, Amber Glass
Class A Volumetric Flasks, Amber Glass
Class A Volumetric Flasks, Amber Glass
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks, Amber Glass are manufactured from durable DURAN™ glass tubing.
Have Questions?
Catalog NumberCapacity (Metric)Standard Taper Joint
FKS11736742500 mL19/26 NS Standard Ground
Catalog number FKS11736742
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Standard Taper Joint:
19/26 NS Standard Ground
  • With NS ground and hollow amber glass stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Certifications/ComplianceDIN EN ISO 1042
Closure ColorBrown
Closure MaterialGlass
ColorBrown
Capacity (Metric)500 mL
MaterialBorosilicate Glass, Duran Glass
Height (Metric)260 mm
Limit of Error±0.25 mL
Standard Taper Joint19/26 NS Standard Ground
Tolerance±0.25 mL
GraduatedYes
ClassA
Unit SizePK