Class A Amber Glass Volumetric Flasks with Poly Stopper
Class A Amber Glass Volumetric Flasks with Poly Stopper
Actual product may vary
Class A Amber Glass Volumetric Flasks with Poly Stopper
Class A Amber Glass Volumetric Flasks with Poly Stopper
Fisherbrand
Class A Amber Glass Volumetric Flasks with Poly Stopper
Allows for precise, safe liquid measurement and handling of light-sensitive materials in the laboratory. Hirschmann Instruments™ Class A Amber Glass Volumetric Flasks with Poly Stopper are manufactured from durable DURAN™ glass tubing.
Have Questions?
Catalog NumberCapacity (Metric)Standard Taper JointTolerance
FKS11736712100 mL14/23 NS Standard Ground±0.1 mL
Catalog number FKS11736712
Price (MYR)
703.00
PK
Add to cart
Capacity (Metric):
100 mL
Standard Taper Joint:
14/23 NS Standard Ground
Tolerance:
±0.1 mL
Price (MYR)
703.00
PK
Add to cart
  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Certifications/ComplianceDIN EN ISO 1042
Closure MaterialPolyethylene
ColorBrown
Capacity (Metric)100 mL
MaterialBorosilicate Glass, Duran Glass
Height (Metric)170 mm
Limit of Error±0.1 mL
Standard Taper Joint14/23 NS Standard Ground
Tolerance±0.1 mL
GraduatedYes
ClassA
Unit SizePK