Traceable™ Electronic Timer Clock: 10-Hour
Traceable™ Electronic Timer Clock: 10-Hour
Actual product may vary
Fisherbrand
Traceable™ Electronic Timer Clock: 10-Hour
One-channel benchtop timer with 1-second resolution and 0.01% accuracy
Have Questions?
Catalog number FKS11725873
Price (MYR)
317.00
EA
Add to cart
Price (MYR)
317.00
EA
Add to cart
Fisher Scientific™ Traceable™ 10-Hour Electronic Timer Clock is a one-channel benchtop timer with 1-second resolution and 0.01% accuracy.
  • Memory recall
  • Full keyboard
  • Timing capacity: 10 hrs.
  • Display: 0.75 in.H LCD, readable from 20 ft.
  • Clock/Stopwatch with AM/PM indicators
  • Alarm: high decibel, 1 minute, silenced automatically or manually
  • Memory reset
  • Dimensions: 2.54L x 6.35W x 7.62cmH (1 × 2.5 × 3 in.)
Specifications
Product TypeBenchtop Timer
Time Range10 hr.
Alarms1 min. Audible/Visible
Accuracy±0.01%
Resolution1 sec.
DisplayLCD
Height (English) Display0.75 in.
Height (Metric) Display1.9 cm
Weight (English)2.5 oz.
Weight (Metric)71 g
For Use With (Application)For general purpose timing applications
Certifications/ComplianceNIST, ISO 17025
Unit SizeEA