Microglass Fiber Filters, Grade 261
Microglass Fiber Filters, Grade 261
Actual product may vary
Fisherbrand
Microglass Fiber Filters, Grade 261
Made with 100% borosilicate micro-glass fibers and are binder free. Fisherbrand™ Microglass Fiber Filters are particularly suitable for micro-filtration because of very fine fibers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS11724083110 mm
FKS1171408390 mm
Catalog number FKS11724083
Price (MYR)
661.00
PK
Add to cart
Diameter (Metric):
110 mm
Price (MYR)
661.00
PK
Add to cart
  • High loading capacity
  • Excellent levels of particles retention (pore size 0.7 to 2.7μm)
  • Chemical resistance to many solvents and reagents
  • Non-hygroscopic and biologically inert
  • HIgh permeability against the passing of air
  • Suitable for use at and up to 500°C
Applications: filtration of industrial water, ligand binding test, proteins filtration, radio-immuno tests, suspended sollids in water (EN872 norm), drinking water filtration, cells collection
Specifications
Basis Weight52 g/m2
Filter TypeMicrofilter
FormatDisc
MaterialBorosilicate Glass
Pore Size1.2 μm
Filtration Time100 sec.
For Use With (Application)Filtration of industrial water, ligand binding test, Proteins filtration, radio-immuno tests, suspended solids in water, drinking water filtration, cells collection
Grade261
Surface TypeNon-hygroscopic
Temperature (Metric) Max500°C
Thickness (English)0.01 in.
Thickness (Metric)0.26 mm
TypeFilter
Diameter (Metric)110 mm
Unit SizePK