Check Order Status
Oxygen Bottles
Actual product may vary
Oxygen Bottles
Individually marked and calibrated oxygen bottles acc. To winkler
Have Questions?
Catalog Number
Volume (Metric)
FKS11712293
200 to 300 mL
Catalog number
FKS11712293
Price (MYR)
1,009.00
PK
Add to cart
Volume (Metric):
200 to 300 mL
Price (MYR)
1,009.00
PK
Add to cart
Acc. to Winkler
White graduation
Precisely adjusted
With labelling square
Consecutively serial-numbered on the bottle and the bevelled glass stopper
Specifications
Volume (Metric)
200 to 300 mL
Height (Metric)
135 mm