Polypropylene 96-Well PCR™ Rack
Polypropylene 96-Well PCR™ Rack
Actual product may vary
Polypropylene 96-Well PCR™ Rack
Polypropylene 96-Well PCR™ Rack
Fisherbrand
Polypropylene 96-Well PCR™ Rack
Ideal for pre- or post-PCR sampling applications. Polypropylene 96-Well PCR™ Rack includes 96 easily identified tube wells. The removable hinged lies flat and the rack is autoclavable. Made of sturdy polypropylene.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS11710344Blue, Green, Orange, Pink, Yellow
FIS03-448-20Blue, Green, Orange, Pink, Yellow
Catalog number FKS11710344
Price (MYR)
382.00
PK
Add to cart
Color:
Blue, Green, Orange, Pink, Yellow
Price (MYR)
382.00
PK
Add to cart
Use with pre- or post-PCR™ sampling applications. Fisherbrand™ Polypropylene 96-Well PCR™ Rack includes 96 easily identified tube wells. The removable hinged lies flat and the rack is autoclavable. Made of sturdy polypropylene.
  • Sturdy polypropylene construction in a variety of colors.
  • Accommodates individual 0.2 mL tubes or strips of 8 or 12 tubes.
  • Removable hinged lid lies flat.
  • Tube wells are easily identified with embossed numbers and letters.
  • Autoclavable and stackable.
Specifications
AutoclavableAutoclavable
IncludesRemovable, hinged lid
Product TypePCR Tube Rack
Closure IncludedYes
AutoclavableAutoclavable
ColorBlue, Green, Orange, Pink, Yellow
HoldsPCR Tubes
MaterialPolypropylene
Positions96
ShapeRectangular
ShelvesNone
StackableYes
SterilityNon-sterile
Dimensions (L x W x H)127 x 101 x 38 mm (5 x 4 x 1.5 in.)
Length (Metric)127 mm
Length (English)5 in.
Width (Metric)101 mm
Width (English)4 in.
Height (Metric)38 mm
Height (English)1.5 in.
Depth (Metric)130 mm
Diameter (Metric) Well5.2 mm
Unit SizePK