HyFlex™ 11-920 Series Blue Nitrile Medium Weight Gloves
HyFlex™ 11-920 Series Blue Nitrile Medium Weight Gloves
Actual product may vary
HyFlex™ 11-920 Series Blue Nitrile Medium Weight Gloves
HyFlex™ 11-920 Series Blue Nitrile Medium Weight Gloves
Fisherbrand
HyFlex™ 11-920 Series Blue Nitrile Medium Weight Gloves
Handle small and medium-sized components lightly coated with oil or lubricant with confidence. Protects skin from oil contamination.
Have Questions?
Catalog NumberSize
FKS117066738
Catalog number FKS11706673
Price (MYR)
-
Size:
8
  • Patented Ansell Grip Technology™.
  • Microscopic channels in the glove coating guide away the oil for secure glove-to-surface contact.
  • Knitted construction
  • Nylon liner
  • Nitrile coating
  • Palm-coated finish
  • Knitwrist cuff
  • Blue color
Specifications
Back MaterialNitrile
Certifications/ComplianceANSI, EN 388 : 3131
Cuff MaterialNitrile
Coating MaterialNitrile
Product TypeMediumweight Gloves
Size8
Style11-920
Glove Cuff StyleKnit
LiningNylon
Palm TypeCoated-palm
Quantity144 Pairs
Resistant ToGood snag, puncture and superior abrasion resistance
Series11-920
Unit SizePAA