Check Order Status
Bright Stainless Steel Rods
Actual product may vary
Fisherbrand
Bright Stainless Steel Rods
An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Bright Stainless Steel Rods are durable and corrosion resistant.
Have Questions?
Catalog Number
Length (Metric)
Diameter (Metric) Inner
FKS11705396
1000 mm
12.5 mm
Catalog number
FKS11705396
Price (MYR)
491.00
EA
Add to cart
Length (Metric):
1000 mm
Diameter (Metric) Inner:
12.5 mm
Price (MYR)
491.00
EA
Add to cart
Durable
Easy-to-clean
Corrosion resistant
Stainless steel
Specifications
Material
Stainless Steel
Length (Metric)
1000 mm
Diameter (Metric) Inner
12.5 mm
Includes
Includes rod, but not the base.
Product Type
Rod (Threaded)
Thread Size
10 mm
Description
Threaded