Spilltray and Drying Rack
Spilltray and Drying Rack
Actual product may vary
Fisherbrand
Spilltray and Drying Rack
Polyethylene, chemical-resistant tray safely contains chemicals and liquids while pipetting, measuring or mixing
Have Questions?
Catalog NumberHolds
FKS117042661.25 L Liquid
Catalog number FKS11704266
Price (MYR)
349.00
EA
Add to cart
Holds:
1.25 L Liquid
Price (MYR)
349.00
EA
Add to cart
Polyethylene, chemical-resistant tray safely contains chemicals and liquids while pipetting, measuring or mixing
  • Holds up to 1.25L (42.5 oz.) of spilled liquid
  • Low profile, plastic grid design cushions and protects glassware and delicate items from breakage and scratches
  • Grid allows faster drying time by permitting air circulation
  • Easy to clean–dishwasher safe
Specifications
Includes1 Tray
Product TypeDrying Rack
AutoclavableNot Autoclavable
ColorBlue/White
Holds1.25 L Liquid
MaterialPlastic
ShapeRectangular
ShelvesNone
StackableNo
SterilityNon-sterile
For Use With (Equipment)Chemicals and Liquids
Dimensions (L x W x H)16.5 x 13 x 0.625 in.
Length (Metric)44 cm
Width (Metric)34 cm
Height (Metric)2.54 cm
Unit SizeEA