Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Actual product may vary
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Fisherbrand
Samco™ Fine Tip Transfer Pipettes
Perform precise dispensing of small drops into small-volume containers using these fine-tip transfer pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityBulb Draw (Metric)Packaging Type
FKS116118391.5 mLNon-sterile1.0 mL
FIS13-711-251.7 mLNon-sterile1.0 mLBulk 400/Pk 8Pk/Cs 3200/Cs
FKS103748505.4 mLNon-sterile3.1 mLBulk 250/Pk 8Pk/Cs 2000/Cs
FIS13-711-265.8 mLNon-sterile3.3 mL500/Pk., 5000/Cs.
FIS13-711-315.8 mLNon-sterile3.7 mLBulk 500/Pk 10Pk/Cs 5000/Cs
Catalog number FKS11611839
Price (MYR)
403.00
PK
Add to cart
Volume (Metric):
1.5 mL
Sterility:
Non-sterile
Bulb Draw (Metric):
1.0 mL
Price (MYR)
403.00
PK
Add to cart
These fine-tip transfer pipettes are ideal for removing separating liquid samples. Designed with a built-in pipette bulb, they allow precise dispensing of small drops into small-volume containers.
  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface – prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
  • Available with fine tip or extended fine tip
Specifications
Bulb Draw (Metric)1.0 mL
DescriptionSamco 1.5mL Extended Fine Tip Transfer Pipets, Small Bulb, Non-sterile, White Box
Diameter (Metric) Outer3.0 mm
MaterialLDPE
SterilityNon-sterile
GraduatedNo
Quantity400/Pk.
Volume (Metric)1.5 mL
Length (Metric)10.4 cm
TypeExtended Fine Tip Transfer Pipet
Unit SizePK