Eutech Handheld Meter Calibration Solution, 480 mL 111.8 mS/cm KCl
Eutech Handheld Meter Calibration Solution, 480 mL 111.8 mS/cm KCl
Eutech Handheld Meter Calibration Solution, 480 mL 111.8 mS/cm KCl
Eutech Handheld Meter Calibration Solution, 480 mL 111.8 mS/cm KCl
Fisherbrand
Eutech Handheld Meter Calibration Solution, 480 mL 111.8 mS/cm KCl
Periodic calibration with accurate standards is recommended for meter
Have Questions?
Catalog NumberQuantity
FKS11594684
Catalog number FKS11594684
Price (MYR)
286.00
Price (MYR)
286.00
EA
Add to cart
Chemical Identifiers
CAS7447-40-7
Specifications
Chemical Name or MaterialInorganic Salts
CAS7447-40-7
Recommended StorageKeep container tightly closed. Keep in cool dark area away from light
OdorOdorless
Density1.0 g/cm³ at 25°C
View more
Periodic calibration with accurate standards is recommended for meter
  • 480 mL 111.8 mS/cm KC calibration solution
  • Solutions packed in 480mL durable plastic bottles
  • Economical and convenient for laboratory use.