Borosilicate Glass Weighing Funnels
Borosilicate Glass Weighing Funnels
Actual product may vary
Fisherbrand
Borosilicate Glass Weighing Funnels
Introduce powder via the tubular stem. Fisherbrand™ Borosilicate Glass Weighing Funnels are ideal for weighing small quantities of powder.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)
FKS1159290210 mL100 mm
FKS115729023 mL70 mm
FKS115829026 mL85 mm
Catalog number FKS11592902
Price (MYR)
123.00
EA
Add to cart
Capacity (Metric):
10 mL
Height (Metric):
100 mm
Price (MYR)
123.00
EA
Add to cart
Ideal for weighing small quantities of powder. Powder can be introduced via the tubular stem.
Specifications
Capacity (Metric)10 mL
MaterialBorosilicate Glass
Height (Metric)100 mm
For Use With (Application)Ideal for weighing small quantities of powder
Product TypeWeighing Funnel
Unit SizeEA