Storage Vials
Storage Vials
Actual product may vary
Storage Vials
Storage Vials
Fisher Catalogue
Storage Vials
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ColorClosure SizeQuantity
FKS1158374040 mLClear20-400100 Vials
B7999-12 mLClear, Clear8-425100 Vials
B7815-20
also known as NSC GROUP B7815-20
White20-400100 Caps
B7995-13White, Clear13-425100 Septa
B7995-15White, Clear15-425100 Septa
FKS10445082White24 mm100
Catalog number FKS11583740
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
40 mL
Color:
Clear
Closure Size:
20-400
Quantity:
100 Vials
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.
Unassembled convenience kits saves time during sample preparation.
  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description24-400 Screw Vial, 40 mL
Diameter (Metric) Inner28 mm
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass
Volume (Metric)40 mL
Base StyleFlat Bottom
Volume (Metric) Total40 mL
Height (Metric)95 mm
Quantity100 Vials
TypeScrew Thread Vial
Closure Size20-400
Marking SpotNo
Unit SizePK