Red Rubber Tubing, Heavy Wall
Red Rubber Tubing, Heavy Wall
Actual product may vary
Fisherbrand
Red Rubber Tubing, Heavy Wall
Highly resilient, heavy-wall red rubber tubing. Fisherbrand™ Red Rubber Tubing is suitable for vacuum use and the transfer of liquids, gases, and dilute acids.
Have Questions?
Catalog NumberDiameter (Metric) InnerDiameter (Metric) OuterThickness (Metric)
FKS115775625 mm13 mm4 mm
Catalog number FKS11577562
Price (MYR)
589.00
EA
Add to cart
Diameter (Metric) Inner:
5 mm
Diameter (Metric) Outer:
13 mm
Thickness (Metric):
4 mm
Price (MYR)
589.00
EA
Add to cart
  • Higher resilience than most other polymers
  • Temperature range: -40°C to 70°C
  • Light and flexible, for transferring liquids, gases and dilute acids
  • Heavy wall recommended for vacuum use
  • Not suitable for use with petroleum oils and hydrocarbon solvents (see neoprene tubing)
Specifications
Diameter (Metric) Inner5 mm
MaterialRubber
Diameter (Metric) Outer13 mm
Overall Length (Metric)10 m
Thickness (Metric)4 mm
For Use With (Application)For transferring liquids, gases and dilute acids
ColorRed
Unit SizeEA