Adhesive PCR Plate Seals
Adhesive PCR Plate Seals
Actual product may vary
Adhesive PCR Plate Seals
Thermo Scientific™ ABgene Adhesive PCR Plate Seals provide an effective and convenient sealing method for PCR applications.
Have Questions?
Catalog NumberQuantity
FKS11570274100 sheets
Catalog number FKS11570274
Price (MYR)
-
Quantity:
100 sheets
Highlights
  • Clear for sample inspection
  • Strong adhesive prevents evaporation during thermal cycling or sample storage
  • No sticky residue on the plate when the seal is peeled off
  • End tabs for easy seal application and removal
  • Recommended for use in conjunction with a cycler that has a screw-down or clip-down lid
Specifications
ColorClear
DescriptionPeelable adhesive PCR film
No. per Pack100
Quantity100 sheets
Product TypeFilm, Seal, or Applicator
AdhesiveAdhesive Clear Seal
Temperature Range (Metric)-20°C to +120°C
Plate Compatibility96/384-well Plate
Unit SizePK