Check Order Status
Borosilicate Glass Graduated Cylinder
Actual product may vary
Fisherbrand
Borosilicate Glass Graduated Cylinder
Measure liquids with blue ceramic scale graduated glass cylindars. The Fisherbrand™ borosilicate glass graduated cylindawith hexagonal base and spout
Have Questions?
Catalog Number
Capacity (Metric)
Graduations
FKS11547702
100 mL
1 mL
Catalog number
FKS11547702
Price (MYR)
-
Capacity (Metric):
100 mL
Graduations:
1 mL
DIN 12680, BS 604, ISO 4788
Graduated in blue ceramic
Borosilicate Glass
Hexagonal base
Spouted
Individual reorder code on each product
Specifications
Material
Borosilicate Glass
Certifications/Compliance
DIN 12680, BS 604, ISO 4788
Class
B
Capacity (Metric)
100 mL
Scale
Blue Ceramic
Graduations
1 mL