Polystyrene Semi Micro Cuvettes for Visible Wavelengths
Polystyrene Semi Micro Cuvettes for Visible Wavelengths
Actual product may vary
Fisherbrand
Polystyrene Semi Micro Cuvettes for Visible Wavelengths
Use these cuvettes for more constant sample temperatures during photometric measurements. Fisherbrand™ Polystyrene Semi Micro Cuvettes for Visible Wavelengths feature excellent optical transmission range and small wall thickness for increased heat transfer.
Have Questions?
Catalog NumberCapacity (Metric)
FKS115476921.6 mL
Catalog number FKS11547692
Price (MYR)
62.00
PK
Add to cart
Capacity (Metric):
1.6 mL
Price (MYR)
62.00
PK
Add to cart
Semi-micro cuvette, disposable polystyrene.
  • Capacity 1.6mL
  • Transparency approximately 90% between 400nm and 800nm
  • Range 336nm to 850nm
  • Lightpath 10mm
  • Dimensions 12. 5mm x 45mm
Specifications
TypeSemi-micro Cuvette
DisposableSingle-use
Capacity (Metric)1.6 mL
MaterialPolystyrene
Path Length10 mm
Wavelength Range400 to 800 nm
Height (Metric)45 mm
Width (Metric)12.5 mm
Unit SizePK