Check Order Status
Stainless Steel Micro Chattaway Spatulas
Actual product may vary
Fisherbrand
Stainless Steel Micro Chattaway Spatulas
Scrapes and lifts materials from tiny bottles or flasks. Fisherbrand™ Stainless Steel Chattaway Spatulas are non-magnetic with a bright finish.
Have Questions?
Catalog Number
Length (Metric)
FKS11543482
150 mm
Catalog number
FKS11543482
Price (MYR)
145.00
PK
Add to cart
Length (Metric):
150 mm
Price (MYR)
145.00
PK
Add to cart
Manufactured from Stainless Steel
Bent and rounded end scrapes and lifts materials from bottles and flasks
Fits into tight flasks and bottles with necks ≥ 4mm diameter
Specifications
Quantity
5 Pk.
Product Type
Chattaway Spatula
Width (Metric)
4 mm
Diameter (Metric) Outer
1.6 mm (Barrel)
Dimensions (L x W)
150 x 4 mm
Material
Stainless Steel
Length (Metric)
150 mm