Polystyrene Macro Cuvettes for Visible Wavelengths
Polystyrene Macro Cuvettes for Visible Wavelengths
Actual product may vary
Fisherbrand
Polystyrene Macro Cuvettes for Visible Wavelengths
Use these cuvettes for more constant sample temperatures during photometric measurements. Fisherbrand™ Polystyrene Macro Cuvettes for Visible Wavelengths feature excellent optical transmission range and small wall thickness for increased heat transfer.
Have Questions?
Catalog NumberCapacity (Metric)
FKS115376924 mL
Catalog number FKS11537692
Price (MYR)
62.00
PK
Add to cart
Capacity (Metric):
4 mL
Price (MYR)
62.00
PK
Add to cart
  • Very low variation of extinction values
  • Excellent optical transmission range
  • Cavity-sorted production
  • Transparency Approximately 90% off between 400nm and 800nm
Specifications
TypeMacro Cuvette
DisposableSingle-use
Capacity (Metric)4 mL
MaterialPolystyrene, Polystyrene
Quantity100/Pk.
Path Length10 mm
Wavelength Range400 to 800 nm
Height (Metric)45 mm
Width (Metric)12.5 mm
Unit SizePK