FB70155 Pump
FB70155 Pump
Actual product may vary
FB70155 Pump
Oil-free piston pump. Fisherbrand™ FB70155 Pump designed for general duty laboratory applications requiring only rough vacuum or moderate pressure.
Have Questions?
Catalog NumberDepth (Metric)
FKS11533485194 mm
Catalog number FKS11533485
Price (MYR)
3,597.00
EA
Add to cart
Depth (Metric):
194 mm
Price (MYR)
3,597.00
EA
Add to cart
  • Tough, lightweight, plastic housing with integral carrying handle
  • Water jet or aspirator replacement for general duties
  • Lightweight, compact, portable, standard-duty operation
  • Not recommended for organic, basic and acidic vapors
  • DN6 hose nozzle for 6mm ID vacuum hose
Specifications
Max. Vacuum292 mbar
Electrical Requirements230 V 50 Hz motor with Schuko and UK power leads
Dimensions (L x W x H)20.5 x 12 x 21.0 cm (8.7 x 4.7 x 8.2 in.)
Depth (Metric)194 mm
ModelFB70155
Weight (Metric)2.3 kg
Overall Height (Metric)191 mm
Overall Width (Metric)114 mm
Pump TypePump
Air Displacement9.21 L/min.
ApplicationsGas Sampling
Maximum Pressure2.3 bar
Unit SizeEA