Leakproof HDPE Wide-Mouth Bottles
Leakproof HDPE Wide-Mouth Bottles
Actual product may vary
Leakproof HDPE Wide-Mouth Bottles
Leakproof HDPE Wide-Mouth Bottles
Leakproof HDPE Wide-Mouth Bottles
Fisher Catalogue
Leakproof HDPE Wide-Mouth Bottles
Rigid, translucent bottles.
Have Questions?
Catalog NumberCapacity (Metric)Neck Finish
FKS115280621000 mL63mm
Catalog number FKS11528062
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Neck Finish:
63mm
Rigid, translucent bottles
  • Leakproof†
  • Higher temperature-use rating than LDPE—up to 120°C (248°F) for short periods and 110°C (230°F) for continuous use
  • Brittleness temperature at -100°C (-148°F)
  • Bottles may be radiation, EtO or chemically sterilized
Specifications
Capacity32 oz., 1000 mL
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorTranslucent
MaterialHDPE
SterilityNon-sterile
Neck Finish63mm
Certifications/ComplianceISO 13485:2003; BSI:FM 653694; QMI-SAI:File No.1606319 and 1606321
Closure IncludedYes
AutoclavableNot Autoclavable
MouthWide
Product TypeWide Mouth Bottle
Closure MaterialPolypropylene
Closure Size63 mm
Closure TypeScrew Cap
Unit SizePK