Check Order Status
Polyethylene, Narrow Neck Jerry Can
Actual product may vary
Fisherbrand
Polyethylene, Narrow Neck Jerry Can
Closure Material:
Polyethylene
Have Questions?
Catalog number
FKS11517443
Price (MYR)
71.00
EA
Add to cart
Price (MYR)
71.00
EA
Add to cart
Translucent polyethylene Jerrycan with polyethylene closure
Specifications
Capacity (Metric)
5000 mL
Closure Color
Translucent
Closure Liner
Polyethylene
Closure Material
Polyethylene
Closure Type
Tamper
Diameter (Metric) Inner Neck
189 mm
Material
Polyethylene
Neck Style
Narrow
No. per Pack
1 Ea.