Grade 913 Lens Cleaning Tissue
Grade 913 Lens Cleaning Tissue
Actual product may vary
Fisherbrand
Grade 913 Lens Cleaning Tissue
Used for cleaning of optical lenses.
Have Questions?
Catalog NumberDimensions (L x W)Quantity
FKS11517362135 x 95 mm50 Sheets
Catalog number FKS11517362
Price (MYR)
31.00
PK
Add to cart
Dimensions (L x W):
135 x 95 mm
Quantity:
50 Sheets
Price (MYR)
31.00
PK
Add to cart
  • Very thin paper
  • Made with 100% Manilla fibers
  • Not fluffy
  • Weight: 12g/m2
Application
  • Cleaning of various optical lens
  • Cleaning of optical fibers filaments
Specifications
Product TypeLens Cleaning Tissue
Dimensions (L x W)135 x 95 mm
For Use With (Application)Cleaning of various optical lens (objective, microscopes binoculars, magnifying Glass, Glasses, etc.), cleaning of optical fibers filaments
MaterialManilla Fibers
Quantity50 Sheets
Unit SizePK