Stainless Steel Chattaway Spatulas
Stainless Steel Chattaway Spatulas
Actual product may vary
Fisherbrand
Stainless Steel Chattaway Spatulas
Scrapes and lifts materials from bottles or flasks. Fisherbrand™ Stainless Steel Chattaway Spatulas are non-magnetic with a bright finish.
Have Questions?
Catalog NumberLength (Metric)
FKS11513472150 mm
Catalog number FKS11513472
Price (MYR)
210.00
PK
Add to cart
Length (Metric):
150 mm
Price (MYR)
210.00
PK
Add to cart
  • Manufactured from Stainless Steel
  • Bent and rounded end scrapes and lifts materials from bottles and flasks
Specifications
Quantity5 Pk.
Product TypeChattaway Spatula
Width (Metric)10 mm
Diameter (Metric) Outer3.6 mm
Dimensions (L x W)150 x 10 mm
MaterialStainless Steel
Length (Metric)150 mm
Unit SizePK