Check Order Status
Dissolved Oxygen Membrane Kit
Actual product may vary
Dissolved Oxygen Membrane Kit
For use with Fisherbrand™ accumet™ Benchtop Dissolved Oxygen Meters
Have Questions?
Catalog Number
Probe Type
FKS11510264
DO
Catalog number
FKS11510264
Price (MYR)
3,168.00
EA
Add to cart
Probe Type:
DO
Price (MYR)
3,168.00
EA
Add to cart
For use with Fisherbrand™ accumet™ Benchtop Dissolved Oxygen Meters
Includes six membrane caps, polishing disk and electrolyte filling solution
Specifications
For Use With (Equipment)
Accumet Benchtop DO Meter
Type
Membrane Kit
Includes
Six membrane caps, polishing disk and electrolyte filling solution
Probe Type
DO