Polypropylene Microcentrifuge Tubes
Polypropylene Microcentrifuge Tubes
Actual product may vary
Fisherbrand
Polypropylene Microcentrifuge Tubes
Use these tubes for high speed centrifugation applications. Fisherbrand™ Polypropylene Microcentrifuge Tubes feature a frosted writing surface on closure and side of tube (except for natural tubes) which allows for easy and convenient sample identification.
Have Questions?
Catalog NumberColorCapacity (Metric)
FKS11508232Natural0.5 mL
Catalog number FKS11508232
Price (MYR)
171.00
PK
Add to cart
Color:
Natural
Capacity (Metric):
0.5 mL
Price (MYR)
171.00
PK
Add to cart
  • Graduated polypropylene tubes
  • Suitable for withstanding the stress of high speed centrifugation up to 20,000xg
  • One-piece construction incorporates a snug fitting and reliable cap and the bottom is reinforced for added protection against leakage
  • Autoclavable
Specifications
Closure MaterialPolypropylene
AutoclavableAutoclavable
Capacity (Metric)0.5 mL
ColorNatural
MaterialPolypropylene
For Use With (Equipment)Microcentrifuge
No. per Pack1000/Pk.
Packaging1000/Pk.
Max. RCF20,000 x g
Unit SizePK