Check Order Status
Thermographic Measurements Temperature Strip
Actual product may vary
Thermographic Measurements Temperature Strip
Have Questions?
Catalog Number
Temperature Range (Metric)
FKS11502513
121°C to 160°C
Catalog number
FKS11502513
Price (MYR)
-
Temperature Range (Metric):
121°C to 160°C
The standard 8 level range covers 40°C to 260°C for general purpose temperature monitoring.
Specifications
For Use With (Application)
For general purpose temperature monitoring
Product Type
Temperature Strip
Max. Temperature (Metric)
160°C
Min. Temperature (Metric)
121°C
Temperature Range (Metric)
121°C to 160°C