Spectra/Por™ Cellulose Regenerated Tubing Membrane
Spectra/Por™ Cellulose Regenerated Tubing Membrane
Actual product may vary
Fisherbrand
Spectra/Por™ Cellulose Regenerated Tubing Membrane
For molecular separation, protein binding studies, electrolyte removal and desalting
Have Questions?
Catalog NumberMWCOLength (Metric)Capacity (Metric)
FKS1149585912 to 14 kDa15 m46 mL/cm
Catalog number FKS11495859
Price (MYR)
6,155.00
EA
Add to cart
MWCO:
12 to 14 kDa
Length (Metric):
15 m
Capacity (Metric):
46 mL/cm
Price (MYR)
6,155.00
EA
Add to cart
Pure regenerated natural cellulose membranes with 1kDa to 50kDa MWCO. Transparent and flexible except Spectra/Por™ 5. Low molecular weight compounds in aqueous solutions diffuse through the membrane, higher molecular weights, such as proteins and bacteria, are retained by the membrane. Convenient dispenser boxes protect unused membranes. Glycerol (where used) preserves membrane and must be removed before use. Soak membrane for three hours in deionised water, then rinse well with running water. Spectra/Por™ 5 is reinforced with a layer of porous paper, to give a thick, rigid membrane for use in high shear environments.
  • Spectra/Por™ 1, 2, 3, 4, 5, supplied dry with glycerol
  • Spectra/Por™ 6, pre-wetted supplied in 0.1% sodium azide
  • Spectra/Por™ 7, pre-treated to reduce trace contaminants supplied in 0.1% sodium azide
For molecular separation, protein binding studies, electrolyte removal and desalting.
Specifications
Product TypeRegenerated Tubing Membrane
MWCO12 to 14 kDa
MaterialCellulose
Length (Metric)15 m
Capacity (Metric)46 mL/cm
ColorTransparent
Temperature Range (Metric)60°C
Quantity1 Each
For Use With (Application)For molecular separation, protein binding studies, electrolyte removal and desalting
Unit SizeEA