Grade 115 Cellulose Very Slowly Filtering Qualitative Filter Paper
Grade 115 Cellulose Very Slowly Filtering Qualitative Filter Paper
Actual product may vary
Fisherbrand
Grade 115 Cellulose Very Slowly Filtering Qualitative Filter Paper
Fisherbrand™ Grade 115 Cellulose Very Slowly Filtering Qualitative Filter Paper are made of cellulose fibers.
Have Questions?
Catalog NumberFormatDiameter (Metric)
FKS11455198Disc125 mm
Catalog number FKS11455198
Price (MYR)
160.00
PK
Add to cart
Format:
Disc
Diameter (Metric):
125 mm
Price (MYR)
160.00
PK
Add to cart
  • Pure filter paper grades made of cellulose fibers with alpha content of nearly 100% according precise norms
  • Ash content for these grades is approximatively 0.06 %
Specifications
DescriptionAsh Content: 0.06%
FormatDisc
MaterialCellulose
Particle Retention2 to 3 μm
Pore Size2 to 3 μm
For Use With (Application)For extremely difficult filtration, Barium Sulphate, Metastannic Acid, Cuprous Oxide
Grade115
Quantity100/Pk.
Thickness (Metric)0.14 mm
Diameter (Metric)125 mm
Unit SizePK