Amber Borosilicate Glass Class A Volumetric Flask with Stopper
Amber Borosilicate Glass Class A Volumetric Flask with Stopper
Actual product may vary
Fisherbrand
Amber Borosilicate Glass Class A Volumetric Flask with Stopper
Amber Borosilicate Glass Volumetric Flask includes interchangeable polyethylene stopper.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeTolerance
FKS1139943325 mL10/19±0.04 mL
FKS113594431000 mL24/29±0.4 mL
FKS11339443250 mL14/23±0.15 mL
FKS11349443500 mL19/24±0.25 mL
FKS1130944350 mL12/21±0.06 mL
FKS11319443100 mL12/21±0.01 mL
Catalog number FKS11399433
Price (MYR)
572.00
PK
Add to cart
Capacity (Metric):
25 mL
Closure Size:
10/19
Tolerance:
±0.04 mL
Price (MYR)
572.00
PK
Add to cart
Amber Borosilicate Glass Volumetric Flask includes interchangeable polyethylene stopper.
  • With one mark graduation and interchangeable polyethylene stopper.
  • The details on each flask include confirmation of compliance to International Standards as well as an individual re-order code.
  • The Fisherbrand 'Batch Certificate of Compliance' is downloadable for this item from the 'Documentation' tab
Specifications
Certifications/ComplianceISO 1042, DIN 12664
Closure MaterialPolyethylene
ColorAmber
Capacity (Metric)25 mL
MaterialBorosilicate Glass
Height (Metric)110 mm
Tolerance±0.04 mL
GraduatedYes
ClassA
Closure Size10/19
Unit SizePK