Check Order Status
Kimble Chase™ Soda Lime Glass Culture Test Tubes
Actual product may vary
Fisherbrand
Kimble Chase™ Soda Lime Glass Culture Test Tubes
Use Soda Glass Cell Culture Test Tubes to analyze cell culture material that are critical to diagnoses and experimentation.
Have Questions?
Catalog Number
Length (Metric)
Diameter (Metric)
FKS11387554
100 mm
13 mm
Catalog number
FKS11387554
Price (MYR)
198.00
PK
Add to cart
Length (Metric):
100 mm
Diameter (Metric):
13 mm
Price (MYR)
198.00
PK
Add to cart
Soda lime glass
Round bottom
X250
Specifications
Design Type
Round-bottom
Description
Test Tube
Length (Metric)
100 mm
Material
Soda Lime Glass
Quantity
250 Pack
Autoclavable
Autoclavable
Diameter (Metric)
13 mm