Non-Indicating FTA™ Micro Card
Non-Indicating FTA™ Micro Card
Actual product may vary
Fisherbrand
Non-Indicating FTA™ Micro Card
Collect, stabilize, process, transport, and archive blood samples. Non-Indicating FTA™ Micro Card are used for DNA analysis by direct amplification, standard amplification with rinsing (punch-in method) and extraction of DNA from the card.
Have Questions?
Catalog NumberFor Use With (Application)
FKS11370304Forensics, Transgenic Identification, HLA Typing/Transfusion Medicine
Catalog number FKS11370304
Price (MYR)
5,830.00
PK
Add to cart
For Use With (Application):
Forensics, Transgenic Identification, HLA Typing/Transfusion Medicine
Price (MYR)
5,830.00
PK
Add to cart
FTA technology consists of two distinct chemistries, both of which have the ability to lyse cells on contact, denature proteins, and protect DNA from degradation. FTA chemistry contains chemical denaturants and a free radical scavenger. With FTA, the DNA remains tightly bound while proteins and inhibitors are washed from the matrix.
  • One sample area per card
  • Improves process efficiency by enabling direct amplification for STR analysis
  • Non-indicating cards for colored samples, like blood
  • Immediate stabilization and protection of nucleic acids due to FTA technology
  • Different card options to support fast manual or automated processing
  • Long term archiving of samples at room temperature due to FTA card technology
Specifications
FormatCard
Sample TypeBlood
For Use With (Application)Forensics, Transgenic Identification, HLA Typing/Transfusion Medicine
No. of Samples1
Product TypeNon-Indicating FTA Micro Card
Quantity125 μL
Sample Volume125μL
Unit SizePK